Assay Details
Target Gene Details
Entrez Gene ID: | 7182 |
Gene Name: | nuclear receptor subfamily 2 group C member 2 |
Gene Aliases: |
TAK1, TR4 |
Location: |
Chr.3:14947584-15049279 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR2C2 | NM_001291694.1 | NP_001278623.1 | ||
| NM_003298.4 | NP_003289.2 | |||
| XM_011534058.2 | XP_011532360.1 | |||
| XM_011534059.2 | XP_011532361.1 | |||
| XM_011534061.2 | XP_011532363.1 | |||
| XM_011534063.2 | XP_011532365.1 | |||
| XM_011534064.2 | XP_011532366.1 | |||
| XM_011534065.2 | XP_011532367.1 | |||
| XM_011534066.2 | XP_011532368.1 | |||
| XM_017007118.1 | XP_016862607.1 | |||
| XM_017007119.1 | XP_016862608.1 | |||
| AK290590.1 | ||||
| BC051670.1 | AAH51670.1 | |||
| L27586.1 | AAA21474.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955319 | Chr.3:14982594 - 14991593 on Build GRCh38 | Duplication |
|
| esv2759130 | Chr.3:14869763 - 15121196 on Build GRCh38 | Gain |
|
| nsv834620 | Chr.3:14912162 - 15093316 on Build GRCh38 | Gain |
|
| esv3595377 | Chr.3:14912760 - 14994553 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map