Assay Details
Target Gene Details
Entrez Gene ID: | 55768 |
Gene Name: | N-glycanase 1 |
Gene Aliases: |
CDDG, CDG1V, PNG1, PNGase |
Location: |
Chr.3:25718944-25790039 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NGLY1 | NM_001145294.1 | NP_001138766.1 | ||
| AK296047.1 | ||||
| DC329733.1 |
Target Gene Details
Entrez Gene ID: | 54995 |
Gene Name: | 3-oxoacyl-ACP synthase, mitochondrial |
Gene Aliases: |
FASN2D, KASI, KS |
Location: |
Chr.3:25783661-25794534 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXSM | XM_006713216.3 | 1 | 1060 | XP_006713279.1 |
| XM_006713217.3 | 1 | 1060 | XP_006713280.1 | |
| XM_017006713.1 | 1 | 1060 | XP_016862202.1 | |
| XM_017006714.1 | 1 | 1060 | XP_016862203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893676 | Chr.3:24984204 - 26577498 on Build GRCh38 | Loss |
|
| nsv3736 | Chr.3:25752343 - 25785188 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map