Assay Details
Target Gene Details
Entrez Gene ID: | 285195 |
Gene Name: | solute carrier family 9 member A9 |
Gene Aliases: |
AUTS16, NHE9 |
Location: |
Chr.3:143265222-143848531 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A9 | NM_173653.3 | NP_775924.1 | ||
| XM_011512703.2 | XP_011511005.1 | |||
| XM_017006202.1 | XP_016861691.1 | |||
| XM_017006203.1 | XP_016861692.1 | |||
| AB075486.1 | BAE45746.1 | |||
| AB089794.1 | BAD69592.1 | |||
| AK092932.1 | BAC04005.1 | |||
| AK301816.1 | ||||
| AK315107.1 | ||||
| AL832304.1 | CAI46158.1 | |||
| AY254100.1 | AAP80573.1 | |||
| BC035779.1 | AAH35779.1 |
Target Gene Details
Entrez Gene ID: | 100885796 |
Gene Name: | SLC9A9 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:143342246-143347071 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A9-AS1 | NR_048544.1 | |||
| DB338547.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759184 | Chr.3:143273855 - 143567137 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map