Assay Details
Target Gene Details
Entrez Gene ID: | 100873979 |
Gene Name: | RBMS3 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.3:29264194-29280939 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBMS3-AS3 | NR_109804.1 | |||
| BG201526.1 | ||||
| BM840572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010530 | Chr.3:29245678 - 30074618 on Build GRCh38 | Gain |
|
| nsv590026 | Chr.3:29246160 - 29579113 on Build GRCh38 | Gain |
|
| esv2759137 | Chr.3:29142404 - 29653997 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76669169] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map