Assay Details
Target Gene Details
Entrez Gene ID: | 6189 |
Gene Name: | ribosomal protein S3A |
Gene Aliases: |
FTE1, MFTL, S3A |
Location: |
Chr.4:151099573-151104652 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS3A | NM_001006.4 | NP_000997.1 | ||
| NM_001267699.1 | NP_001254628.1 | |||
| AK129825.1 | ||||
| AK291075.1 | ||||
| AK296121.1 | ||||
| AK308947.1 | ||||
| AK311788.1 | ||||
| BC001708.1 | AAH01708.1 | |||
| BC004981.1 | AAH04981.1 | |||
| BQ055367.1 | ||||
| L13802.1 | AAA35682.1 | |||
| M77234.1 | AAA60290.1 | |||
| M84711.1 | AAA58487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv595690 | Chr.4:150867803 - 151188400 on Build GRCh38 | Gain |
|
| nsv964104 | Chr.4:151100977 - 151107733 on Build GRCh38 | Duplication |
|
| esv3602674 | Chr.4:151094056 - 151193747 on Build GRCh38 | Gain |
|
| dgv5436n100 | Chr.4:150743286 - 151150724 on Build GRCh38 | Gain |
|
| nsv1028814 | Chr.4:150874356 - 152285445 on Build GRCh38 | Gain |
|
| nsv526976 | Chr.4:150754667 - 151133921 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map