Assay Details
Target Gene Details
Entrez Gene ID: | 10699 |
Gene Name: | corin, serine peptidase |
Gene Aliases: |
ATC2, CRN, Lrp4, PEE5, TMPRSS10 |
Location: |
Chr.4:47593998-47838106 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORIN | NM_001278585.1 | NP_001265514.1 | ||
| NM_006587.3 | NP_006578.2 | |||
| AF113248.1 | AAF21966.1 | |||
| AF133845.1 | AAD31850.1 | |||
| AK302816.1 | ||||
| AK304038.1 | ||||
| BC110451.2 | AAI10452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829923 | Chr.4:47434122 - 47676426 on Build GRCh38 | Gain |
|
| dgv5202n100 | Chr.4:47486414 - 47904818 on Build GRCh38 | Gain |
|
| nsv1003341 | Chr.4:47605279 - 47649975 on Build GRCh38 | Gain |
|
| nsv594118 | Chr.4:47558369 - 47915713 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map