Assay Details
Target Gene Details
Entrez Gene ID: | 84803 |
Gene Name: | glycerol-3-phosphate acyltransferase 3 |
Gene Aliases: |
AGPAT 10, AGPAT10, AGPAT8, AGPAT9, HMFN0839, LPAAT-theta, MAG1 |
Location: |
Chr.4:83535634-83605875 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPAT3 | NM_001256421.1 | NP_001243350.1 | ||
| NM_001256422.1 | NP_001243351.1 | |||
| NM_032717.4 | NP_116106.2 | |||
| XM_011532384.2 | XP_011530686.1 | |||
| XM_017008780.1 | XP_016864269.1 | |||
| XM_017008781.1 | XP_016864270.1 | |||
| AK055749.1 | BAB71002.1 | |||
| AK223543.1 | BAD97263.1 | |||
| AY358100.1 | AAQ88467.1 | |||
| BC090956.1 | AAH90956.1 | |||
| DA393801.1 | ||||
| DA673378.1 | ||||
| DQ324782.1 | ||||
| DQ345298.1 | ABC70186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829994 | Chr.4:83445146 - 83637163 on Build GRCh38 | Loss |
|
| nsv829995 | Chr.4:83500542 - 83557306 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map