Assay Details
Target Gene Details
Entrez Gene ID: | 643226 |
Gene Name: | glutaredoxin and cysteine rich domain containing 2 |
Gene Aliases: |
DFNB101 |
Location: |
Chr.5:145858538-145931673 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRXCR2 | NM_001080516.1 | NP_001073985.1 | ||
| XM_017009708.1 | XP_016865197.1 | |||
| DN831963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3570480 | Chr.5:145827318 - 145873808 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs184037544] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map