Assay Details
Target Gene Details
Entrez Gene ID: | 285600 |
Gene Name: | KIAA0825 |
Gene Aliases: |
C5orf36 |
Location: |
Chr.5:94150851-94618604 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0825 | NM_001145678.1 | NP_001139150.1 | ||
| NM_173665.2 | NP_775936.1 | |||
| XM_006714588.2 | XP_006714651.1 | |||
| XM_011543324.2 | XP_011541626.1 | |||
| XM_011543325.2 | XP_011541627.1 | |||
| XM_011543328.2 | XP_011541630.1 | |||
| XM_011543329.2 | XP_011541631.1 | |||
| XM_011543330.2 | XP_011541632.1 | |||
| XM_011543331.1 | XP_011541633.1 | |||
| XM_011543332.1 | XP_011541634.1 | |||
| XM_011543333.2 | XP_011541635.1 | |||
| XM_011543334.2 | XP_011541636.1 | |||
| XM_017009373.1 | XP_016864862.1 | |||
| XM_017009374.1 | XP_016864863.1 | |||
| XM_017009375.1 | XP_016864864.1 | |||
| XM_017009376.1 | XP_016864865.1 | |||
| XM_017009377.1 | XP_016864866.1 | |||
| AK308195.1 | ||||
| BC035515.1 | AAH35515.1 | |||
| BX648338.1 |
Target Gene Details
Entrez Gene ID: | 84250 |
Gene Name: | SMC5-SMC6 complex localization factor 1 |
Gene Aliases: |
ANKRD32, BRCTD1, BRCTx |
Location: |
Chr.5:94613477-94697733 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLF1 | XM_017009979.1 | 1 | 1672 | XP_016865468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598952 | Chr.5:94488246 - 94621281 on Build GRCh38 | Loss |
|
| esv2759354 | Chr.5:94429500 - 94706496 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map