Assay Details
Target Gene Details
Entrez Gene ID: | 92255 |
Gene Name: | LMBR1 domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.5:36098550-36151961 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMBRD2 | XM_017010024.1 | 19 | 7786 | XP_016865513.1 |
| AK094621.1 | 1 | 3080 | ||
| BX537605.1 | 1 | 3501 | ||
| BX647926.1 | 1 | 1608 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604707 | Chr.5:35513484 - 36501574 on Build GRCh38 | Gain |
|
| esv32711 | Chr.5:36047695 - 36146064 on Build GRCh38 | Gain |
|
| nsv830262 | Chr.5:36074209 - 36293770 on Build GRCh38 | Gain |
|

Genomic Map