Assay Details
Target Gene Details
Entrez Gene ID: | 92255 |
Gene Name: | LMBR1 domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.5:36098550-36151961 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMBRD2 | NM_001007527.1 | NP_001007528.1 | ||
| XM_011514162.2 | XP_011512464.1 | |||
| XM_017010024.1 | XP_016865513.1 | |||
| AK091295.1 | ||||
| AK300433.1 | ||||
| AL137370.1 | CAB70714.1 | |||
| BC110506.2 | AAI10507.1 | |||
| BX647340.1 | ||||
| CR749399.1 | CAH18245.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3604707 | Chr.5:35513484 - 36501574 on Build GRCh38 | Gain |
|
| esv32711 | Chr.5:36047695 - 36146064 on Build GRCh38 | Gain |
|
| nsv830262 | Chr.5:36074209 - 36293770 on Build GRCh38 | Gain |
|

Genomic Map