Assay Details
Target Gene Details
Entrez Gene ID: | 578 |
Gene Name: | BCL2 antagonist/killer 1 |
Gene Aliases: |
BAK, BAK-LIKE, BCL2L7, CDN1 |
Location: |
Chr.6:33572546-33580293 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAK1 | NM_001188.3 | NP_001179.1 | ||
| XM_011514779.2 | XP_011513081.1 | |||
| XM_011514780.1 | XP_011513082.1 | |||
| AK091807.1 | ||||
| AK292093.1 | ||||
| AK293547.1 | ||||
| AK303422.1 | ||||
| CB142221.1 | ||||
| CR627020.1 | 1 | 1563 | CAI46254.1 | |
| U16811.1 | AAA74466.1 | |||
| U23765.1 | AAA93066.1 | |||
| X84213.1 | CAA58997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608630 | Chr.6:33439052 - 34076935 on Build GRCh38 | Gain |
|
| nsv602833 | Chr.6:33567103 - 33620370 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map