Assay Details
Target Gene Details
Entrez Gene ID: | 135138 |
Gene Name: | PARK2 coregulated |
Gene Aliases: |
GLUP, HAK005771, PARK2CRG |
Location: |
Chr.6:162726870-163315492 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv605159 | Chr.6:162379124 - 163058475 on Build GRCh38 | Loss |
|
| esv2759489 | Chr.6:162419890 - 162812229 on Build GRCh38 | Gain |
|
| nsv1015507 | Chr.6:162444113 - 162913747 on Build GRCh38 | Gain |
|
| nsv605180 | Chr.6:162673891 - 162791267 on Build GRCh38 | Loss |
|
| esv3571505 | Chr.6:161937230 - 162847872 on Build GRCh38 | Loss |
|
| nsv830857 | Chr.6:162643779 - 162824781 on Build GRCh38 | Loss |
|
| nsv1026490 | Chr.6:162712401 - 163284676 on Build GRCh38 | Gain |
|
| dgv6176n100 | Chr.6:162161146 - 162762531 on Build GRCh38 | Gain |
|
| nsv517562 | Chr.6:162060428 - 162828613 on Build GRCh38 | Gain+Loss |
|
| nsv1018573 | Chr.6:162604329 - 162751777 on Build GRCh38 | Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map