Assay Details
Target Gene Details
Entrez Gene ID: | 387319 |
Gene Name: | histone cluster 1, H2a, pseudogene 1 |
Gene Aliases: |
H2AFSP, bA317E16.5 |
Location: |
Chr.6:25732433-25732861 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H2APS1 | NR_045125.1 | |||
| DB441866.1 |
Target Gene Details
Entrez Gene ID: | 6568 |
Gene Name: | solute carrier family 17 member 1 |
Gene Aliases: |
NAPI-1, NPT-1, NPT1 |
Location: |
Chr.6:25723744-25832108 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A1 | XM_017011201.1 | XP_016866690.1 |
More Information
Set Membership: |
|

Genomic Map