Assay Details
Target Gene Details
Entrez Gene ID: | 26 |
Gene Name: | amine oxidase, copper containing 1 |
Gene Aliases: |
ABP, ABP1, DAO, DAO1, KAO |
Location: |
Chr.7:150824875-150861291 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AOC1 | NM_001091.3 | NP_001082.2 | ||
| NM_001272072.1 | NP_001259001.1 | |||
| XM_017011944.1 | XP_016867433.1 | |||
| XM_017011945.1 | XP_016867434.1 | |||
| AK092514.1 | ||||
| AK296638.1 | ||||
| AK307278.1 | ||||
| AK314806.1 | ||||
| BC014093.2 | AAH14093.1 | |||
| BX648159.1 | ||||
| M55602.1 | AAA58358.1 | |||
| U11862.1 | AAC50270.1 | |||
| U11863.1 | AAB60381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6754n100 | Chr.7:150850025 - 150877464 on Build GRCh38 | Loss |
|
| nsv1030109 | Chr.7:150615828 - 151003922 on Build GRCh38 | Gain |
|
| nsv608990 | Chr.7:150264185 - 151010400 on Build GRCh38 | Gain |
|
| esv2759574 | Chr.7:150841733 - 150966609 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map