Assay Details
Target Gene Details
Entrez Gene ID: | 79989 |
Gene Name: | tetratricopeptide repeat domain 26 |
Gene Aliases: |
DYF13, IFT56, dyf-13 |
Location: |
Chr.7:139133744-139191986 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC26 | NM_001144920.2 | NP_001138392.1 | ||
| NM_001144923.2 | NP_001138395.1 | |||
| NM_001287512.1 | NP_001274441.1 | |||
| NM_001287513.1 | NP_001274442.1 | |||
| NM_001318333.1 | NP_001305262.1 | |||
| NM_001321740.1 | NP_001308669.1 | |||
| NM_001321741.1 | NP_001308670.1 | |||
| NM_001321742.1 | NP_001308671.1 | |||
| NM_024926.3 | NP_079202.2 | |||
| AK022633.1 | BAB14143.1 | |||
| AK293444.1 | ||||
| AK295092.1 | ||||
| AK299132.1 | ||||
| AK300807.1 | ||||
| AL137393.1 | CAB70721.2 | |||
| BC013912.1 | AAH13912.1 | |||
| BC126331.1 | ||||
| BC130339.1 | ||||
| BP315162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831155 | Chr.7:138953647 - 139144186 on Build GRCh38 | Loss |
|
| esv2759567 | Chr.7:139048951 - 139242593 on Build GRCh38 | Gain |
|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map