Assay Details
Target Gene Details
Entrez Gene ID: | 8795 |
Gene Name: | TNF receptor superfamily member 10b |
Gene Aliases: |
CD262, DR5, KILLER, KILLER/DR5, TRAIL-R2, TRAILR2, TRICK2, TRICK2A, TRICK2B, TRICKB, ZTNFR9 |
Location: |
Chr.8:23020133-23069187 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF10B | NM_003842.4 | NP_003833.4 | ||
| NM_147187.2 | NP_671716.2 | |||
| NR_027140.1 | ||||
| XM_017013944.1 | XP_016869433.1 | |||
| AB451272.1 | ||||
| AB451399.1 | ||||
| AF012535.1 | AAB67103.1 | |||
| AF012628.1 | AAB67109.1 | |||
| AF016266.1 | AAB81180.1 | |||
| AF016268.1 | AAC01565.1 | |||
| AF016849.1 | AAC51778.1 | |||
| AF018657.1 | AAB70577.1 | |||
| AF018658.1 | AAB70578.1 | |||
| AF020501.1 | AAB71412.1 | |||
| AF022386.1 | AAB71949.1 | |||
| AF153687.1 | AAF75587.1 | |||
| AF192548.1 | AAF07175.1 | |||
| AK291216.1 | ||||
| AK296128.1 | ||||
| AK298602.1 | ||||
| AY358277.1 | AAQ88644.1 | |||
| BC001281.1 | AAH01281.1 | |||
| BX537617.1 | CAD97801.1 | |||
| BX538104.1 | CAD98017.1 | |||
| BX538179.1 | CAD98053.1 | |||
| CR541898.1 | CAG46696.1 | |||
| CX870630.1 | ||||
| FJ200482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027718 | Chr.8:22805950 - 23258334 on Build GRCh38 | Gain |
|
| nsv524231 | Chr.8:23003729 - 23028617 on Build GRCh38 | Loss |
|
| nsv1025672 | Chr.8:22868571 - 23130480 on Build GRCh38 | Gain |
|
| esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
|
| nsv1017570 | Chr.8:22969562 - 23544783 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map