Assay Details
Target Gene Details
Entrez Gene ID: | 6542 |
Gene Name: | solute carrier family 7 member 2 |
Gene Aliases: |
ATRC2, CAT2, HCAT2 |
Location: |
Chr.8:17494349-17570568 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A2 | XM_017013746.1 | XP_016869235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610680 | Chr.8:17483518 - 17505730 on Build GRCh38 | Loss |
|
| nsv831246 | Chr.8:17430363 - 17621085 on Build GRCh38 | Gain |
|
| esv3616489 | Chr.8:17490020 - 17737549 on Build GRCh38 | Loss |
|
| nsv1024271 | Chr.8:17418630 - 17540313 on Build GRCh38 | Loss |
|
| esv3616490 | Chr.8:17490532 - 17587102 on Build GRCh38 | Loss |
|
| esv34867 | Chr.8:17488520 - 17725642 on Build GRCh38 | Loss |
|
| nsv516180 | Chr.8:17424902 - 17541640 on Build GRCh38 | Loss |
|
| esv2759600 | Chr.8:17433671 - 17784021 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map