Assay Details
Target Gene Details
Entrez Gene ID: | 1755 |
Gene Name: | deleted in malignant brain tumors 1 |
Gene Aliases: |
GP340, SAG, muclin |
Location: |
Chr.10:122560665-122643736 on Build GRCh38 |
Assay Gene Location: | Within Intron 43 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428240 | Chr.10:122503559 - 122695299 on Build GRCh38 | Gain+Loss |
|
| nsv8731 | Chr.10:122579399 - 122622194 on Build GRCh38 | Gain+Loss |
|
| esv3624781 | Chr.10:122602807 - 122624657 on Build GRCh38 | Loss |
|
| nsv508611 | Chr.10:122562819 - 122630985 on Build GRCh38 | Deletion |
|
| esv3546970 | Chr.10:122592440 - 122623563 on Build GRCh38 | Deletion |
|
| dgv294e212 | Chr.10:122582087 - 122622928 on Build GRCh38 | Loss |
|
| nsv1159773 | Chr.10:122564250 - 122629780 on Build GRCh38 | Duplication |
|
| dgv299e212 | Chr.10:122598537 - 122622928 on Build GRCh38 | Loss |
|
| nsv499308 | Chr.10:122580319 - 122622028 on Build GRCh38 | Loss |
|
| nsv56 | Chr.10:122598443 - 122627790 on Build GRCh38 | Deletion |
|
| nsv7581 | Chr.10:122569936 - 122639333 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map