Assay Details
Target Gene Details
Entrez Gene ID: | 80114 |
Gene Name: | BicC family RNA binding protein 1 |
Gene Aliases: |
BICC, CYSRD |
Location: |
Chr.10:58512970-58831437 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BICC1 | NM_001080512.2 | NP_001073981.1 | ||
| XM_011540185.2 | XP_011538487.1 | |||
| AY726586.1 | ||||
| CF242850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508586 | Chr.10:58516186 - 58621289 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map