Assay Details
Target Gene Details
Entrez Gene ID: | 3983 |
Gene Name: | actin binding LIM protein 1 |
Gene Aliases: |
ABLIM, LIMAB1, LIMATIN, abLIM-1 |
Location: |
Chr.10:114431110-114821643 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Gene Details
Entrez Gene ID: | 101927692 |
Gene Name: | uncharacterized LOC101927692 |
Gene Aliases: |
- |
Location: |
Chr.10:114764788-114779903 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927692 | NR_120628.1 | |||
| AK098198.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038050 | Chr.10:114555350 - 115135954 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map