Assay Details
Target Gene Details
Entrez Gene ID: | 219970 |
Gene Name: | glycine-N-acyltransferase like 2 |
Gene Aliases: |
BXMAS2-10, GATF-B |
Location: |
Chr.11:58834065-58905699 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLYATL2 | XM_011544809.2 | XP_011543111.1 | ||
| XM_017017338.1 | XP_016872827.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23547 | Chr.11:58847198 - 58869038 on Build GRCh38 | Gain+Loss |
|
| nsv1050013 | Chr.11:58627125 - 59453617 on Build GRCh38 | Gain |
|
| dgv71n21 | Chr.11:58786298 - 58864895 on Build GRCh38 | Loss |
|
| dgv139e201 | Chr.11:58845738 - 59113363 on Build GRCh38 | Deletion |
|
| dgv1207n100 | Chr.11:58767187 - 58911023 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map