Assay Details
Target Gene Details
Entrez Gene ID: | 5436 |
Gene Name: | RNA polymerase II subunit G |
Gene Aliases: |
RPB19, RPB7, hRPB19, hsRPB7 |
Location: |
Chr.11:62761539-62766715 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2G | NM_002696.2 | NP_002687.1 | ||
| AK312131.1 | ||||
| BC112162.1 | AAI12163.1 | |||
| BC112164.1 | AAI12165.1 | |||
| BG530809.1 | ||||
| U20659.1 | AAA86500.1 |
More Information
Set Membership: |
|

Genomic Map