Assay Details
Target Gene Details
Entrez Gene ID: | 219972 |
Gene Name: | macrophage expressed 1 |
Gene Aliases: |
MPG1, MPS1, Mpg-1 |
Location: |
Chr.11:59208510-59213021 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPEG1 | NM_001039396.1 | 1 | 3770 | NP_001034485.1 |
| AK024358.1 | 2 | 1379 | ||
| AK074166.1 | 2 | 3673 | ||
| AK095457.1 | 2 | 2823 | ||
| AK097211.1 | 1 | 2033 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050013 | Chr.11:58627125 - 59453617 on Build GRCh38 | Gain |
|
| esv2759830 | Chr.11:58932619 - 59241200 on Build GRCh38 | Gain |
|

Genomic Map