Assay Details
Target Gene Details
Entrez Gene ID: | 100130460 |
Gene Name: | uncharacterized LOC100130460 |
Gene Aliases: |
- |
Location: |
Chr.11:10308313-10430673 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAND1.11 | NR_103765.1 | |||
| EF537579.1 | ||||
| EF537580.1 | ||||
| EF537581.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2518130 | Chr.11:10364109 - 10365527 on Build GRCh38 | Deletion |
|

Genomic Map