Assay Details
Target Gene Details
Entrez Gene ID: | 112937 |
Gene Name: | galactosidase beta 1 like 3 |
Gene Aliases: |
- |
Location: |
Chr.11:134275447-134321364 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLB1L3 | NM_001080407.2 | NP_001073876.2 | ||
| XM_011542566.2 | XP_011540868.1 | |||
| XM_011542567.2 | XP_011540869.1 | |||
| XM_011542568.2 | XP_011540870.1 | |||
| XM_011542569.2 | XP_011540871.1 | |||
| XM_011542570.2 | XP_011540872.1 | |||
| XM_011542571.2 | XP_011540873.1 | |||
| XM_017017155.1 | XP_016872644.1 | |||
| XM_017017156.1 | XP_016872645.1 | |||
| BC011001.2 | AAH11001.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1302n100 | Chr.11:134275705 - 134348093 on Build GRCh38 | Loss |
|
| nsv521581 | Chr.11:134126584 - 134772738 on Build GRCh38 | Gain |
|
| nsv826149 | Chr.11:134277313 - 134278343 on Build GRCh38 | Loss |
|
| esv3628164 | Chr.11:134276952 - 134344689 on Build GRCh38 | Loss |
|
| nsv826148 | Chr.11:134273214 - 134282719 on Build GRCh38 | Gain |
|
| nsv468901 | Chr.11:134266299 - 134340635 on Build GRCh38 | Loss |
|
| nsv556598 | Chr.11:134252356 - 134298912 on Build GRCh38 | Loss |
|
| esv3892114 | Chr.11:134054794 - 134328278 on Build GRCh38 | Gain |
|
| nsv1037294 | Chr.11:134260280 - 134340956 on Build GRCh38 | Loss |
|
| nsv832299 | Chr.11:134170258 - 134346785 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs67473835] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map