Assay Details
Target Gene Details
Entrez Gene ID: | 84747 |
Gene Name: | unc-119 lipid binding chaperone B |
Gene Aliases: |
POC7B |
Location: |
Chr.12:120710435-120723640 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC119B | NM_001080533.2 | NP_001074002.1 | ||
| AK126367.1 | ||||
| DA324219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2658765 | Chr.12:120714331 - 120715701 on Build GRCh38 | Deletion |
|
| nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
|
| esv3630930 | Chr.12:120714386 - 120715610 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map