Assay Details
Target Gene Details
Entrez Gene ID: | 283464 |
Gene Name: | glucoside xylosyltransferase 1 |
Gene Aliases: |
GLT8D3 |
Location: |
Chr.12:42081845-42144871 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GXYLT1 | NM_001099650.1 | NP_001093120.1 | ||
| NM_173601.1 | NP_775872.1 | |||
| XM_017019211.1 | XP_016874700.1 | |||
| AK125141.1 | ||||
| BC030023.1 | AAH30023.1 | |||
| BC039145.1 | AAH39145.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv983314 | Chr.12:42117709 - 42119851 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map