Assay Details
Target Gene Details
Entrez Gene ID: | 8847 |
Gene Name: | deleted in lymphocytic leukemia 2 (non-protein coding) |
Gene Aliases: |
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS |
Location: |
Chr.13:49982552-50125541 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLEU2 | NR_002612.1 | |||
| AY455757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580637 | Chr.13:50006792 - 50829882 on Build GRCh38 | Loss |
|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
| nsv1035151 | Chr.13:49939399 - 50593714 on Build GRCh38 | Loss |
|
| nsv1054415 | Chr.13:50023985 - 50076802 on Build GRCh38 | Loss |
|

Genomic Map