Assay Details
Target Gene Details
Entrez Gene ID: | 55775 |
Gene Name: | tyrosyl-DNA phosphodiesterase 1 |
Gene Aliases: |
- |
Location: |
Chr.14:89954935-90044764 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832854 | Chr.14:89877925 - 90070680 on Build GRCh38 | Gain |
|
| nsv832853 | Chr.14:89818942 - 89990332 on Build GRCh38 | Gain |
|
| nsv1045864 | Chr.14:89882005 - 89968468 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map