Assay Details
Target Gene Details
Entrez Gene ID: | 283554 |
Gene Name: | G protein-coupled receptor 137C |
Gene Aliases: |
TM7SF1L2 |
Location: |
Chr.14:52553148-52637713 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR137C | NM_001099652.1 | NP_001093122.1 | ||
| AK123953.1 | ||||
| AL834372.1 | CAD39035.1 |

Genomic Map