Assay Details
Target Gene Details
Entrez Gene ID: | 64745 |
Gene Name: | methyltransferase like 17 |
Gene Aliases: |
METT11D1 |
Location: |
Chr.14:20989806-20997035 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL17 | NM_001029991.1 | NP_001025162.1 | ||
| NM_022734.2 | NP_073571.1 | |||
| XM_006720236.2 | XP_006720299.1 | |||
| XM_017021601.1 | XP_016877090.1 | |||
| AF321002.1 | AAK08200.1 | |||
| AK024512.1 | BAB14919.1 | |||
| AK303484.1 | ||||
| AK304180.1 | ||||
| AK315999.1 | ||||
| BC005053.1 | AAH05053.1 | |||
| KU178747.1 | ||||
| KU178748.1 | ||||
| KU178749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428293 | Chr.14:20747949 - 21063624 on Build GRCh38 | Gain+Loss |
|
| nsv563847 | Chr.14:20870665 - 21000992 on Build GRCh38 | Loss |
|
| nsv832740 | Chr.14:20863901 - 21076893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map