Assay Details
Target Gene Details
Entrez Gene ID: | 84888 |
Gene Name: | signal peptide peptidase like 2A |
Gene Aliases: |
IMP3, PSL2 |
Location: |
Chr.15:50702266-50765713 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPPL2A | NM_032802.3 | NP_116191.2 | ||
| XM_005254722.3 | XP_005254779.1 | |||
| XM_017022680.1 | XP_016878169.1 | |||
| XM_017022681.1 | XP_016878170.1 | |||
| AJ345028.1 | CAC87789.1 | |||
| AJ420896.1 | CAD13133.1 | |||
| AK027446.1 | BAB55117.1 | |||
| AK075454.1 | BAC11630.1 | |||
| AK315651.1 | ||||
| AY169314.1 | AAO12539.1 | |||
| BC025740.1 | AAH25740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv317n27 | Chr.15:50478671 - 50897510 on Build GRCh38 | Gain |
|
| nsv1047443 | Chr.15:50452584 - 51354826 on Build GRCh38 | Gain |
|
| nsv569398 | Chr.15:50496928 - 51002798 on Build GRCh38 | Loss |
|
| nsv1051400 | Chr.15:50598136 - 50754086 on Build GRCh38 | Gain |
|
| dgv2594n100 | Chr.15:50469213 - 50823338 on Build GRCh38 | Gain |
|
| dgv4526n54 | Chr.15:50582767 - 51475068 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map