Assay Details
Target Gene Details
Entrez Gene ID: | 2923 |
Gene Name: | protein disulfide isomerase family A member 3 |
Gene Aliases: |
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC |
Location: |
Chr.15:43746392-43772606 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDIA3 | NM_005313.4 | NP_005304.3 | ||
| AK075455.1 | ||||
| AK075481.1 | ||||
| AK293249.1 | ||||
| BC014433.1 | AAH14433.1 | |||
| BC036000.1 | ||||
| BC071878.1 | AAH71878.1 | |||
| BG826781.1 | ||||
| D16234.1 | BAA03759.1 | |||
| D83485.1 | BAA11928.1 | |||
| EU668338.1 | ||||
| FJ224330.1 | ||||
| U42068.1 | AAC50331.1 | |||
| Z49835.1 | CAA89996.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636297 | Chr.15:43697364 - 43747461 on Build GRCh38 | Gain |
|
| esv2760028 | Chr.15:43530960 - 43856443 on Build GRCh38 | Gain+Loss |
|
| nsv976928 | Chr.15:43656668 - 43748688 on Build GRCh38 | Duplication |
|
| esv33661 | Chr.15:43724321 - 43772222 on Build GRCh38 | Loss |
|
| esv3569520 | Chr.15:43596064 - 43762536 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map