Assay Details
Target Gene Details
Entrez Gene ID: | 79875 |
Gene Name: | thrombospondin type 1 domain containing 4 |
Gene Aliases: |
ADAMTSL-6, ADAMTSL6, FVSY9334, PRO34005 |
Location: |
Chr.15:71115616-71783383 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THSD4 | NM_024817.2 | NP_079093.2 | ||
| XM_006720692.3 | XP_006720755.1 | |||
| AK131551.1 | BAD18685.1 | |||
| BC140868.1 |
Target Gene Details
Entrez Gene ID: | 101929196 |
Gene Name: | THSD4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:71166812-71189059 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THSD4-AS1 | NR_120348.1 | |||
| NR_120349.1 | ||||
| NR_120350.1 | ||||
| NR_120351.1 | ||||
| DA243226.1 | ||||
| DA296094.1 | ||||
| DA302156.1 | ||||
| DA516711.1 | ||||
| DA798087.1 | ||||
| DN992601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4587n54 | Chr.15:71183376 - 71185709 on Build GRCh38 | Loss |
|
| dgv4590n54 | Chr.15:71183826 - 71185709 on Build GRCh38 | Loss |
|
| nsv469595 | Chr.15:71166185 - 71333967 on Build GRCh38 | Loss |
|
| esv3636851 | Chr.15:71182063 - 71185594 on Build GRCh38 | Loss |
|
| esv3552648 | Chr.15:71181289 - 71185779 on Build GRCh38 | Deletion |
|
| nsv569820 | Chr.15:71181876 - 71185654 on Build GRCh38 | Loss |
|
| esv3581758 | Chr.15:71172593 - 71186817 on Build GRCh38 | Loss |
|
| nsv471251 | Chr.15:71139637 - 71411981 on Build GRCh38 | Gain |
|
| nsv569840 | Chr.15:71184443 - 71185709 on Build GRCh38 | Loss |
|
| esv2749846 | Chr.15:71182072 - 71185585 on Build GRCh38 | Deletion |
|
| esv2670725 | Chr.15:71182071 - 71185597 on Build GRCh38 | Deletion |
|
| nsv457194 | Chr.15:71143283 - 71409321 on Build GRCh38 | Gain |
|
| esv27935 | Chr.15:71182105 - 71185808 on Build GRCh38 | Loss |
|
| nsv1043590 | Chr.15:71183005 - 71591331 on Build GRCh38 | Loss |
|
| nsv1043423 | Chr.15:71112552 - 71337743 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map