Assay Details
Target Gene Details
Entrez Gene ID: | 360226 |
Gene Name: | protease, serine 41 |
Gene Aliases: |
TESSP1 |
Location: |
Chr.16:2789525-2805287 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS41 | XM_011522477.2 | XP_011520779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map