Assay Details
Target Gene Details
Entrez Gene ID: | 9810 |
Gene Name: | ring finger protein 40 |
Gene Aliases: |
BRE1B, RBP95, STARING |
Location: |
Chr.16:30761612-30800231 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF40 | XM_011545997.1 | XP_011544299.1 |
Target Gene Details
Entrez Gene ID: | 23361 |
Gene Name: | zinc finger protein 629 |
Gene Aliases: |
ZNF, ZNF65 |
Location: |
Chr.16:30778449-30787322 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF629 | NM_001080417.1 | 3 | 1308 | NP_001073886.1 |
| XM_005255222.3 | 3 | 1447 | XP_005255279.1 | |
| AB002324.1 | 3 | 1274 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469724 | Chr.16:30621124 - 30828942 on Build GRCh38 | Loss |
|
| nsv833188 | Chr.16:30621125 - 30828942 on Build GRCh38 | Loss |
|
| nsv9438 | Chr.16:30758882 - 30788121 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map