Assay Details
Target Gene Details
Entrez Gene ID: | 8720 |
Gene Name: | membrane bound transcription factor peptidase, site 1 |
Gene Aliases: |
PCSK8, S1P, SKI-1 |
Location: |
Chr.16:84053761-84116943 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBTPS1 | NM_003791.3 | NP_003782.1 | ||
| XM_017023816.1 | XP_016879305.1 | |||
| XM_017023817.1 | XP_016879306.1 | |||
| AK091212.1 | ||||
| AK291773.1 | ||||
| AK299067.1 | ||||
| BC114555.1 | AAI14556.1 | |||
| BC114961.1 | AAI14962.1 | |||
| BM048055.1 | ||||
| D42053.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892924 | Chr.16:84014717 - 84091665 on Build GRCh38 | Gain |
|
| esv3639410 | Chr.16:84055831 - 84075312 on Build GRCh38 | Loss |
|
| nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss |
|
| esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain |
|
| esv3892927 | Chr.16:84058861 - 84081674 on Build GRCh38 | Gain |
|
| nsv1909 | Chr.16:84031468 - 84076703 on Build GRCh38 | Deletion |
|
| dgv546e214 | Chr.16:84011032 - 84095132 on Build GRCh38 | Gain |
|
| nsv573421 | Chr.16:84013110 - 84091665 on Build GRCh38 | Gain |
|
| nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72799281] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map