Assay Details
Target Gene Details
Entrez Gene ID: | 3250 |
Gene Name: | haptoglobin-related protein |
Gene Aliases: |
A-259H10.2, HP |
Location: |
Chr.16:72063226-72077246 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPR | NM_020995.3 | NP_066275.3 | ||
| AV698541.1 | ||||
| CB115435.1 | ||||
| CB147217.1 | ||||
| T71114.1 |
Target Gene Details
Entrez Gene ID: | 54957 |
Gene Name: | thioredoxin like 4B |
Gene Aliases: |
DLP, Dim2 |
Location: |
Chr.16:72044261-72094316 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNL4B | XM_017023377.1 | XP_016878866.1 | ||
| AK308891.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv504e199 | Chr.16:72049451 - 72076250 on Build GRCh38 | Deletion |
|
| nsv1864 | Chr.16:72064069 - 72084058 on Build GRCh38 | Insertion |
|
| nsv436836 | Chr.16:72064307 - 72074191 on Build GRCh38 | Insertion |
|
| dgv1636e59 | Chr.16:72059952 - 72078350 on Build GRCh38 | Duplication |
|
| esv2714683 | Chr.16:71950823 - 72151945 on Build GRCh38 | Deletion |
|
| nsv833273 | Chr.16:71937902 - 72118955 on Build GRCh38 | Loss |
|
| dgv3017n100 | Chr.16:72040933 - 72757730 on Build GRCh38 | Gain |
|
| nsv978167 | Chr.16:72054423 - 72084772 on Build GRCh38 | Duplication |
|
| nsv572951 | Chr.16:72057155 - 72077447 on Build GRCh38 | Gain |
|
| nsv827729 | Chr.16:72049820 - 72076352 on Build GRCh38 | Loss |
|
| dgv531e214 | Chr.16:72060628 - 72077062 on Build GRCh38 | Gain |
|
| nsv428329 | Chr.16:71942441 - 72240448 on Build GRCh38 | Gain |
|
| nsv9456 | Chr.16:72050862 - 72084423 on Build GRCh38 | Gain+Loss |
|
| esv2422047 | Chr.16:72053111 - 72074194 on Build GRCh38 | Duplication |
|
| esv23639 | Chr.16:72054601 - 72078398 on Build GRCh38 | Gain+Loss |
|
| nsv817737 | Chr.16:72054432 - 72074194 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113186879] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map