Assay Details
Target Gene Details
Entrez Gene ID: | 1535 |
Gene Name: | cytochrome b-245 alpha chain |
Gene Aliases: |
p22-PHOX |
Location: |
Chr.16:88643289-88651084 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYBA | NM_000101.3 | NP_000092.2 | ||
| XM_011522905.2 | XP_011521207.1 | |||
| AK097127.1 | ||||
| AK300049.1 | ||||
| AK309030.1 | ||||
| BC006465.1 | AAH06465.1 | |||
| BT006861.1 | AAP35507.1 | |||
| BX109994.1 | ||||
| M21186.1 | AAA90925.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1926 | Chr.16:88626290 - 88660544 on Build GRCh38 | Insertion |
|
| esv3639554 | Chr.16:88641322 - 88654848 on Build GRCh38 | Loss |
|
| nsv833325 | Chr.16:88553738 - 88698194 on Build GRCh38 | Loss |
|
| nsv952070 | Chr.16:88257295 - 88780192 on Build GRCh38 | Deletion |
|
| nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map