Assay Details
Target Gene Details
Entrez Gene ID: | 65264 |
Gene Name: | ubiquitin conjugating enzyme E2 Z |
Gene Aliases: |
HOYS7, USE1 |
Location: |
Chr.17:48908369-48929060 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2Z | NM_023079.4 | NP_075567.2 | ||
| AB025426.1 | BAB87810.1 | |||
| AK023917.1 | BAB14724.1 | |||
| AK024030.1 | BAB14789.1 | |||
| AK091644.1 | ||||
| AK122770.1 | ||||
| AK130303.1 | ||||
| AK296865.1 | ||||
| AL713782.1 | CAD28542.1 | |||
| BC015169.1 | AAH15169.2 | |||
| BC015890.1 | AAH15890.1 | |||
| CR457322.1 | CAG33603.1 | |||
| EF623992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833472 | Chr.17:48895840 - 49078381 on Build GRCh38 | Gain |
|
| nsv482969 | Chr.17:48843507 - 48994527 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73329977] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map