Assay Details
Target Gene Details
Entrez Gene ID: | 100506677 |
Gene Name: | uncharacterized LOC100506677 |
Gene Aliases: |
AA02, AA04, AA05 |
Location: |
Chr.17:33529787-33533760 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AA06 | NR_037584.2 | |||
| AF381171.1 | ||||
| AF381172.1 | ||||
| AF381173.1 | ||||
| AF381174.1 |
Target Gene Details
Entrez Gene ID: | 40 |
Gene Name: | acid sensing ion channel subunit 2 |
Gene Aliases: |
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1 |
Location: |
Chr.17:33013087-34156806 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASIC2 | NM_001094.4 | NP_001085.2 | ||
| AK313043.1 | ||||
| BC075042.2 | AAH75042.1 | |||
| BC075043.2 | AAH75043.1 | |||
| U50352.1 | AAC50432.1 | |||
| U53212.1 | AAC50498.1 | |||
| U57352.1 | AAB49182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521841 | Chr.17:33406743 - 33593135 on Build GRCh38 | Loss |
|
| nsv9539 | Chr.17:33358415 - 33598698 on Build GRCh38 | Loss |
|
| nsv510706 | Chr.17:33437824 - 33596377 on Build GRCh38 | Deletion |
|
| nsv833421 | Chr.17:33520714 - 33663203 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map