Assay Details
Target Gene Details
Entrez Gene ID: | 80174 |
Gene Name: | DBF4 zinc finger B |
Gene Aliases: |
ASKL1, CHIFB, DRF1, ZDBF1B |
Location: |
Chr.17:44708581-44752732 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv960113 | Chr.17:44710452 - 44721769 on Build GRCh38 | Duplication |
|
| nsv833456 | Chr.17:44639788 - 44825873 on Build GRCh38 | Gain+Loss |
|
| esv3640653 | Chr.17:44713885 - 44719081 on Build GRCh38 | Loss |
|
| esv2663784 | Chr.17:44713888 - 44719054 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map