Assay Details
Target Gene Details
Entrez Gene ID: | 6318 |
Gene Name: | serpin family B member 4 |
Gene Aliases: |
LEUPIN, PI11, SCCA-2, SCCA1, SCCA2 |
Location: |
Chr.18:63637259-63644319 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB4 | NM_002974.3 | NP_002965.1 | ||
| NM_175041.1 | NP_778206.1 | |||
| XM_011526138.1 | XP_011524440.1 | |||
| AB046400.1 | BAB40773.1 | |||
| AF461147.1 | AAQ04771.1 | |||
| AK292212.1 | ||||
| AY245780.1 | AAO92271.1 | |||
| AY245782.1 | AAO92273.1 | |||
| BC017401.1 | AAH17401.1 | |||
| U19557.1 | AAA97553.1 | |||
| X89015.1 | CAA61420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2313855 | Chr.18:63635054 - 63653204 on Build GRCh38 | Deletion |
|
| nsv1066552 | Chr.18:63147299 - 64608176 on Build GRCh38 | Gain |
|
| nsv833672 | Chr.18:63620445 - 63772481 on Build GRCh38 | Gain |
|
| dgv621e214 | Chr.18:63635738 - 63656397 on Build GRCh38 | Loss |
|
| dgv994e212 | Chr.18:63635616 - 63653554 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map