Assay Details
Target Gene Details
Entrez Gene ID: | 28513 |
Gene Name: | cadherin 19 |
Gene Aliases: |
CDH7, CDH7L2 |
Location: |
Chr.18:66501187-66604241 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH19 | NM_001271028.1 | NP_001257957.1 | ||
| NM_021153.3 | NP_066976.1 | |||
| NR_073130.1 | ||||
| XM_011525931.2 | XP_011524233.1 | |||
| XM_017025711.1 | XP_016881200.1 | |||
| AF047826.1 | AAC05117.1 | |||
| AJ007607.1 | CAC13126.1 | |||
| AK304000.1 | ||||
| AK315257.1 | ||||
| AY358654.1 | AAQ89017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458104 | Chr.18:66174053 - 66538654 on Build GRCh38 | Loss |
|
| esv2758734 | Chr.18:66338790 - 66523275 on Build GRCh38 | Gain |
|
| nsv577255 | Chr.18:66316916 - 66722766 on Build GRCh38 | Gain |
|
| nsv1067311 | Chr.18:63818011 - 66637685 on Build GRCh38 | Loss |
|
| dgv3381n100 | Chr.18:66413169 - 66674437 on Build GRCh38 | Loss |
|
| dgv3380n100 | Chr.18:66321439 - 66726779 on Build GRCh38 | Gain |
|
| nsv577257 | Chr.18:66503580 - 66585104 on Build GRCh38 | Loss |
|
| nsv577157 | Chr.18:65715200 - 68002467 on Build GRCh38 | Loss |
|
| esv3642869 | Chr.18:66414787 - 66668426 on Build GRCh38 | Loss |
|
| dgv3382n100 | Chr.18:66506724 - 66615886 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map