Assay Details
Target Gene Details
Entrez Gene ID: | 390858 |
Gene Name: | O-acyltransferase like, pseudogene |
Gene Aliases: |
ACYL3P |
Location: |
Chr.18:59035679-59053214 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OACYLP | NR_024021.2 | |||
| DV080257.1 | ||||
| DV080258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833664 | Chr.18:58947109 - 59093282 on Build GRCh38 | Loss |
|

Genomic Map