Assay Details
Target Gene Details
Entrez Gene ID: | 3594 |
Gene Name: | interleukin 12 receptor subunit beta 1 |
Gene Aliases: |
CD212, IL-12R-BETA1, IL12RB, IMD30 |
Location: |
Chr.19:18058994-18099027 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL12RB1 | NM_001290024.1 | NP_001276953.1 | ||
| XM_006722741.3 | XP_006722804.2 | |||
| XM_011527966.2 | XP_011526268.1 | |||
| XM_011527967.2 | XP_011526269.1 | |||
| XM_011527968.2 | XP_011526270.1 | |||
| XM_011527969.2 | XP_011526271.1 | |||
| XM_011527970.2 | XP_011526272.1 | |||
| XM_011527971.2 | XP_011526273.1 | |||
| XM_011527972.2 | XP_011526274.1 | |||
| XM_011527975.2 | XP_011526277.1 | |||
| XM_011527976.2 | XP_011526278.1 | |||
| XM_011527977.2 | XP_011526279.1 | |||
| BX647221.1 |
Target Gene Details
Entrez Gene ID: | 23031 |
Gene Name: | microtubule associated serine/threonine kinase 3 |
Gene Aliases: |
- |
Location: |
Chr.19:18097764-18151692 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAST3 | NM_015016.1 | NP_055831.1 | ||
| XM_005259824.2 | XP_005259881.1 | |||
| XM_005259825.2 | XP_005259882.1 | |||
| XM_011527823.1 | XP_011526125.1 | |||
| XM_017026512.1 | XP_016882001.1 | |||
| XM_017026513.1 | XP_016882002.1 | |||
| AB011133.1 | BAA25487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470130 | Chr.19:17936474 - 18123777 on Build GRCh38 | Loss |
|
| nsv833773 | Chr.19:18042161 - 18192178 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map