Assay Details
Target Gene Details
Entrez Gene ID: | 388550 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 22, pseudogene |
Gene Aliases: |
- |
Location: |
Chr.19:44537058-44556917 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM22P | NR_027754.2 | |||
| AK123850.1 | ||||
| AK302997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062812 | Chr.19:44538275 - 44711525 on Build GRCh38 | Gain |
|
| nsv483005 | Chr.19:44530751 - 44705087 on Build GRCh38 | Loss |
|

Genomic Map