Assay Details
Target Gene Details
Entrez Gene ID: | 54020 |
Gene Name: | solute carrier family 37 member 1 |
Gene Aliases: |
G3PP |
Location: |
Chr.21:42499632-42581440 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC37A1 | NM_018964.3 | NP_061837.3 | ||
| AJ269529.2 | CAB87248.1 | |||
| AJ277912.1 | CAB91985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522001 | Chr.21:42421796 - 42827928 on Build GRCh38 | Gain |
|
| nsv834103 | Chr.21:42456611 - 42612638 on Build GRCh38 | Gain |
|
| nsv587633 | Chr.21:42485217 - 42631745 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map