Assay Details
Target Gene Details
Entrez Gene ID: | 54456 |
Gene Name: | Mov10 RISC complex RNA helicase like 1 |
Gene Aliases: |
CHAMP, DJ402G11.8 |
Location: |
Chr.22:50090006-50161705 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834236 | Chr.22:50094391 - 50143393 on Build GRCh38 | Loss |
|
| dgv582n27 | Chr.22:50024802 - 50157413 on Build GRCh38 | Gain |
|
| nsv1067062 | Chr.22:50089389 - 50130477 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map